Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGAAAACTTTCTGGGTAGTTGG[C/T]GCGTATGTCTGTGTGCATAGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601437 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGRT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCGRT - Fc fragment of IgG receptor and transporter | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136019.2 | Intron | NP_001129491.1 | ||||
NM_004107.4 | Intron | NP_004098.1 | ||||
XM_005258657.1 | Intron | XP_005258714.1 | ||||
XM_011526627.2 | Intron | XP_011524929.1 |
RCN3 - reticulocalbin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |