Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTAGGGAAGGCGTTATCTCCC[A/C]TAACCAGAAGCTCCAAGGTAAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601026 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP2A1 - adaptor related protein complex 2 alpha 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014203.2 | Intron | NP_055018.2 | ||||
NM_130787.2 | Intron | NP_570603.2 | ||||
XM_011526556.2 | Intron | XP_011524858.1 | ||||
XM_011526557.2 | Intron | XP_011524859.1 |