Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606362 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACPT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACPT - acid phosphatase, testicular | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C19orf48 - chromosome 19 open reading frame 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290149.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277078.1 | |
NM_001290150.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277079.1 | |
NM_001290151.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277080.1 | |
NM_001290152.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277081.1 | |
NM_001290153.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277082.1 | |
NM_001290154.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277083.1 | |
NM_001290155.1 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_001277084.1 | |
NM_032712.2 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_116101.1 | |
NM_199249.2 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_954857.1 | |
NM_199250.2 | 1177 | Silent Mutation | ACC,ACT | T,T 97 | NP_954858.1 |
SNORD88A - small nucleolar RNA, C/D box 88A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD88B - small nucleolar RNA, C/D box 88B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD88C - small nucleolar RNA, C/D box 88C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |