Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACGAGTTTAGCTTCTTTGTGT[C/G]ATCCATTCTCTGCTCAAATGCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605643 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK5 - kallikrein related peptidase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077491.1 | Intron | NP_001070959.1 | ||||
NM_001077492.1 | Intron | NP_001070960.1 | ||||
NM_012427.4 | Intron | NP_036559.1 | ||||
XM_011526702.1 | Intron | XP_011525004.1 | ||||
XM_011526703.2 | Intron | XP_011525005.1 |