Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGTGTGCCTCTTCCAAAGACT[A/G]TATGCCTGGACAAGGGAGCCTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF71 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF71 - zinc finger protein 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021216.4 | 742 | Intron | NP_067039.1 | |||
XM_011527193.2 | 742 | Intron | XP_011525495.1 | |||
XM_017027078.1 | 742 | Intron | XP_016882567.1 | |||
XM_017027079.1 | 742 | UTR 5 | XP_016882568.1 | |||
XM_017027080.1 | 742 | Intron | XP_016882569.1 |