Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGGGTGGGGGGGGTGACATCT[G/T]CAGGCTTCCCTGCTCTGTCCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPATCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPATCH1 - G-patch domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR88 - WD repeat domain 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173479.3 | Intron | NP_775750.3 | ||||
XM_011526449.2 | Intron | XP_011524751.1 | ||||
XM_011526450.2 | Intron | XP_011524752.1 | ||||
XM_017026282.1 | Intron | XP_016881771.1 | ||||
XM_017026283.1 | Intron | XP_016881772.1 |