Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCCCAGCCTGCACCTGTGCAC[A/G]TCCTCCTGCTGAGCCTGACGCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FFAR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FFAR2 - free fatty acid receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005306.2 | 136 | Missense Mutation | ATC,GTC | I,V 46 | NP_005297.1 | |
XM_017026709.1 | 136 | Missense Mutation | ATC,GTC | I,V 46 | XP_016882198.1 | |
XM_017026710.1 | 136 | Missense Mutation | ATC,GTC | I,V 46 | XP_016882199.1 | |
XM_017026711.1 | 136 | Missense Mutation | ATC,GTC | I,V 46 | XP_016882200.1 |