Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACCCAAATAAACCTCTTAATG[C/T]GTTTGTTAAAATTAGTTTGGACATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603289 MIM: 130610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAPK3 - death associated protein kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EEF2 - eukaryotic translation elongation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001961.3 | 2988 | UTR 3 | NP_001952.1 |
SNORD37 - small nucleolar RNA, C/D box 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |