Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGGGTCCCACCGGGTGCCTGC[A/G]CTGTCTTGGGATCTCTGGATACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609765 MIM: 607990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM4B - lysine demethylase 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015015.2 | Intron | NP_055830.1 | ||||
XM_005259521.3 | Intron | XP_005259578.2 | ||||
XM_011527814.2 | Intron | XP_011526116.1 | ||||
XM_011527816.2 | Intron | XP_011526118.1 | ||||
XM_011527817.2 | Intron | XP_011526119.1 | ||||
XM_011527818.2 | Intron | XP_011526120.1 | ||||
XM_011527819.1 | Intron | XP_011526121.1 | ||||
XM_011527820.1 | Intron | XP_011526122.1 | ||||
XM_011527821.1 | Intron | XP_011526123.1 | ||||
XM_011527822.1 | Intron | XP_011526124.1 | ||||
XM_017026503.1 | Intron | XP_016881992.1 | ||||
XM_017026504.1 | Intron | XP_016881993.1 | ||||
XM_017026505.1 | Intron | XP_016881994.1 |
UHRF1 - ubiquitin like with PHD and ring finger domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |