Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCAGTCGGTCTCCAGTGTAGAGC[A/G]AGATAACAGATGAGTGGATGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602895 MIM: 608066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAFB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAFB - scaffold attachment factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201338.1 | Intron | NP_001188267.1 | ||||
NM_001201339.1 | Intron | NP_001188268.1 | ||||
NM_001201340.1 | Intron | NP_001188269.1 | ||||
NM_001320571.1 | Intron | NP_001307500.1 | ||||
NM_001320572.1 | Intron | NP_001307501.1 | ||||
NM_002967.3 | Intron | NP_002958.2 | ||||
XM_017027114.1 | Intron | XP_016882603.1 | ||||
XM_017027115.1 | Intron | XP_016882604.1 | ||||
XM_017027116.1 | Intron | XP_016882605.1 | ||||
XM_017027117.1 | Intron | XP_016882606.1 | ||||
XM_017027118.1 | Intron | XP_016882607.1 |
SAFB2 - scaffold attachment factor B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |