Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAAAAGCTCAACGATGAAGAAA[C/G]AGGAACATGCCCAGGTCAGGTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED16 - mediator complex subunit 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
R3HDM4 - R3H domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138774.3 | 1141 | UTR 3 | NP_620129.2 | |||
XM_011528416.2 | 1141 | Intron | XP_011526718.1 |
RNU6-9 - RNA, U6 small nuclear 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |