Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGTCATTGTTCAGTCCAAAGTA[C/T]TTAATATGATTCTTTTCTCTTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF562 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF562 - zinc finger protein 562 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130031.1 | 4304 | UTR 3 | NP_001123503.1 | |||
NM_001130032.1 | 4304 | UTR 3 | NP_001123504.1 | |||
NM_001300885.1 | 4304 | UTR 3 | NP_001287814.1 | |||
NM_017656.3 | 4304 | UTR 3 | NP_060126.2 | |||
XM_005259941.3 | 4304 | UTR 3 | XP_005259998.1 | |||
XM_005259944.3 | 4304 | UTR 3 | XP_005260001.1 | |||
XM_005259951.3 | 4304 | UTR 3 | XP_005260008.1 | |||
XM_005259952.3 | 4304 | UTR 3 | XP_005260009.1 | |||
XM_011528100.2 | 4304 | UTR 3 | XP_011526402.1 | |||
XM_011528102.2 | 4304 | UTR 3 | XP_011526404.1 | |||
XM_017026898.1 | 4304 | UTR 3 | XP_016882387.1 | |||
XM_017026899.1 | 4304 | UTR 3 | XP_016882388.1 |