Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGGACACTCAGATACCATTAGG[A/G]TGGACTGGGACCCCAGCCTTTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146738 MIM: 600173 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSL3 - insulin like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JAK3 - Janus kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000215.3 | 4768 | UTR 3 | NP_000206.2 | |||
XM_005259896.3 | 4768 | UTR 3 | XP_005259953.1 | |||
XM_011527990.1 | 4768 | Intron | XP_011526292.1 | |||
XM_011527991.2 | 4768 | Intron | XP_011526293.1 |