Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGGGCTGTGGTGAGAGTTATTA[C/T]TCAGGAGACAGACTATGCACAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607368 MIM: 611601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK15 - potassium two pore domain channel subfamily K member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022358.3 | Intron | NP_071753.2 | ||||
XM_017028003.1 | Intron | XP_016883492.1 |
KCNK15-AS1 - KCNK15 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIMS4 - regulating synaptic membrane exocytosis 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |