Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTGGGGAGATAGACTCTGAGA[A/G]GGCCCCAAGAGAAGCAGCTGACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614272 MIM: 610484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASTKD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FASTKD5 - FAST kinase domains 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LZTS3 - leucine zipper tumor suppressor family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282533.1 | 4617 | UTR 3 | NP_001269462.1 | |||
XM_005260890.3 | 4617 | UTR 3 | XP_005260947.1 | |||
XM_005260892.1 | 4617 | UTR 3 | XP_005260949.1 | |||
XM_005260893.2 | 4617 | UTR 3 | XP_005260950.1 | |||
XM_011529407.2 | 4617 | UTR 3 | XP_011527709.1 | |||
XM_011529408.2 | 4617 | UTR 3 | XP_011527710.1 | |||
XM_011529409.2 | 4617 | UTR 3 | XP_011527711.1 |
UBOX5 - U-box domain containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |