Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTCATTGCTCTAAGACCAAGC[A/C]TAAGTTTGATCTTTTGTGTTCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PANK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PANK2 - pantothenate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324191.1 | Intron | NP_001311120.1 | ||||
NM_001324192.1 | Intron | NP_001311121.1 | ||||
NM_001324193.1 | Intron | NP_001311122.1 | ||||
NM_024960.5 | Intron | NP_079236.3 | ||||
NM_153638.3 | Intron | NP_705902.2 | ||||
NM_153640.3 | Intron | NP_705904.1 | ||||
XM_005260835.3 | Intron | XP_005260892.1 | ||||
XM_005260836.4 | Intron | XP_005260893.3 | ||||
XM_011529364.2 | Intron | XP_011527666.1 | ||||
XM_011529365.2 | Intron | XP_011527667.1 | ||||
XM_017028077.1 | Intron | XP_016883566.1 | ||||
XM_017028078.1 | Intron | XP_016883567.1 | ||||
XM_017028079.1 | Intron | XP_016883568.1 |