Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCCCTGGCATTCCACCAGGCCT[A/G]ACAAAGGGGCCATGCGGTGGGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616023 MIM: 147450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCAF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCAF4 - SR-related CTD associated factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145444.1 | 3129 | Silent Mutation | GTC,GTT | V,V 901 | NP_001138916.1 | |
NM_001145445.1 | 3129 | Silent Mutation | GTC,GTT | V,V 894 | NP_001138917.1 | |
NM_020706.2 | 3129 | Silent Mutation | GTC,GTT | V,V 916 | NP_065757.1 | |
XM_005261017.2 | 3129 | Silent Mutation | GTC,GTT | V,V 857 | XP_005261074.1 | |
XM_006724035.2 | 3129 | Silent Mutation | GTC,GTT | V,V 912 | XP_006724098.1 | |
XM_006724036.2 | 3129 | Silent Mutation | GTC,GTT | V,V 890 | XP_006724099.1 | |
XM_017028415.1 | 3129 | Silent Mutation | GTC,GTT | V,V 915 | XP_016883904.1 | |
XM_017028416.1 | 3129 | Silent Mutation | GTC,GTT | V,V 893 | XP_016883905.1 | |
XM_017028417.1 | 3129 | Silent Mutation | GTC,GTT | V,V 835 | XP_016883906.1 |
SOD1 - superoxide dismutase 1, soluble | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |