Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGTATACATTTATGTTTGTTAC[C/T]AAGCTGATTATAACAATGCAGCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRWD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRWD1 - bromodomain and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMG1 - proteasome assembly chaperone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261824.1 | Intron | NP_001248753.1 | ||||
NM_001320795.1 | Intron | NP_001307724.1 | ||||
NM_003720.3 | Intron | NP_003711.1 | ||||
NM_203433.2 | Intron | NP_982257.1 | ||||
XM_017028488.1 | Intron | XP_016883977.1 |