Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACTGGTCCTAAAAAATAATAAC[A/C]ATAATAATAATAATTAGAGAATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PATZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PATZ1 - POZ/BTB and AT hook containing zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014323.2 | 3304 | UTR 3 | NP_055138.2 | |||
NM_032050.1 | 3304 | UTR 3 | NP_114439.1 | |||
NM_032051.1 | 3304 | Intron | NP_114440.1 | |||
NM_032052.1 | 3304 | UTR 3 | NP_114441.1 |
PIK3IP1-AS1 - PIK3IP1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |