Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACCAGCTACACAAGCTGACGCAG[A/G]AGAGCTTGGAATGACAAAACTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605970 MIM: 605971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFPL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFPL3 - ret finger protein like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098535.1 | 10 | Intron | NP_001092005.1 | |||
NM_006604.2 | 10 | Intron | NP_006595.1 | |||
XM_017028534.1 | 10 | Intron | XP_016884023.1 |
RFPL3S - RFPL3 antisense | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC5A4 - solute carrier family 5 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014227.2 | 10 | Intron | NP_055042.1 | |||
XM_006724308.3 | 10 | UTR 5 | XP_006724371.1 | |||
XM_011530342.2 | 10 | UTR 5 | XP_011528644.1 | |||
XM_011530343.2 | 10 | Intron | XP_011528645.1 | |||
XM_011530344.2 | 10 | Intron | XP_011528646.1 | |||
XM_017028920.1 | 10 | UTR 5 | XP_016884409.1 |