Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTAGGTGGCGGTGGGCGTGGTG[C/G]CGGCCTTCAGGGGGTCCCTGCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602911 | ||||||||||||||||||||
Literature Links: |
CACNG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNG2 - calcium voltage-gated channel auxiliary subunit gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006078.3 | 708 | Missense Mutation | GCC,GGC | A,G 281 | NP_006069.1 | |
XM_017028531.1 | 708 | Missense Mutation | GCC,GGC | A,G 195 | XP_016884020.1 |