Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACAGCGGCCAAGGTACATGAGG[A/C]TGTAGCTTATCGCCCTTTTAAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606004 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGA1 - golgi associated, gamma adaptin ear containing, ARF binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001560.2 | 104 | Intron | NP_001001560.1 | |||
NM_001172687.1 | 104 | Intron | NP_001166158.1 | |||
NM_001172688.1 | 104 | Intron | NP_001166159.1 | |||
NM_013365.4 | 104 | Intron | NP_037497.1 | |||
XM_005261517.3 | 104 | UTR 5 | XP_005261574.1 | |||
XM_005261518.1 | 104 | Intron | XP_005261575.1 | |||
XM_005261520.1 | 104 | Intron | XP_005261577.1 | |||
XM_005261522.1 | 104 | Intron | XP_005261579.1 | |||
XM_006724229.1 | 104 | Intron | XP_006724292.1 | |||
XM_011530122.1 | 104 | UTR 5 | XP_011528424.1 | |||
XM_011530123.1 | 104 | Intron | XP_011528425.1 | |||
XM_011530124.1 | 104 | UTR 5 | XP_011528426.1 | |||
XM_017028760.1 | 104 | UTR 5 | XP_016884249.1 | |||
XM_017028761.1 | 104 | Intron | XP_016884250.1 | |||
XM_017028762.1 | 104 | UTR 5 | XP_016884251.1 | |||
XM_017028763.1 | 104 | Intron | XP_016884252.1 | |||
XM_017028764.1 | 104 | Intron | XP_016884253.1 | |||
XM_017028765.1 | 104 | Intron | XP_016884254.1 |