Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGACAAGAGCAGACGACTCCTGA[C/T]TCTCCACCTAGAATCAGAGTGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPO - translocator protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000714.5 | Intron | NP_000705.2 | ||||
NM_001256530.1 | Intron | NP_001243459.1 | ||||
NM_001256531.1 | Intron | NP_001243460.1 |
TTLL12 - tubulin tyrosine ligase like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |