Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGAATACTGCCTGGCCTATCGC[A/G]AATACATGTGTGTGCAGTTATTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300456 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNB3 - cyclin B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033031.2 | 254 | UTR 5 | NP_149020.2 | |||
NM_033670.2 | 254 | UTR 5 | NP_391990.1 | |||
XM_017029913.1 | 254 | UTR 5 | XP_016885402.1 | |||
XM_017029914.1 | 254 | UTR 5 | XP_016885403.1 | |||
XM_017029915.1 | 254 | UTR 5 | XP_016885404.1 | |||
XM_017029916.1 | 254 | UTR 5 | XP_016885405.1 | |||
XM_017029917.1 | 254 | UTR 5 | XP_016885406.1 | |||
XM_017029918.1 | 254 | UTR 5 | XP_016885407.1 |