Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAGCCTTATAATTTTTCAGGGA[C/G]TGATTCACTTACTTTAGTGTTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 400010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX3Y PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX3Y - DEAD-box helicase 3, Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122665.3 | 1731 | Missense Mutation | ACT,AGT | T,S 438 | NP_001116137.1 | |
NM_001302552.2 | 1731 | Intron | NP_001289481.1 | |||
NM_001324195.1 | 1731 | Intron | NP_001311124.1 | |||
NM_004660.4 | 1731 | Intron | NP_004651.2 |