Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAAGAAATATATAAATAGTAC[C/T]CTTCCCAATGATTCAGAGAATATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616979 | ||||||||||||||||||||
Literature Links: |
DTHD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DTHD1 - death domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136536.4 | 999 | Intron | NP_001130008.2 | |||
NM_001170700.2 | 999 | Silent Mutation | ACC,ACT | T,T 153 | NP_001164171.1 | |
XM_006714014.3 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_006714077.1 | |
XM_011513693.2 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_011511995.1 | |
XM_011513694.2 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_011511996.1 | |
XM_011513695.2 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_011511997.1 | |
XM_011513696.2 | 999 | Intron | XP_011511998.1 | |||
XM_017008191.1 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_016863680.1 | |
XM_017008192.1 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_016863681.1 | |
XM_017008193.1 | 999 | Silent Mutation | ACC,ACT | T,T 278 | XP_016863682.1 |