Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGATGACCTCACACAGGATATCA[C/T]GCTAACCAAATTTTTATGCTGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM114A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM114A1 - family with sequence similarity 114 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138389.2 | Intron | NP_612398.2 | ||||
XM_005262672.2 | Intron | XP_005262729.1 | ||||
XM_006714033.2 | Intron | XP_006714096.1 | ||||
XM_017008838.1 | Intron | XP_016864327.1 |
MIR574 - microRNA 574 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |