Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAAAAAAAAAAAAAAAAGGGTA[C/T]TAAGAGGTCAAAGTGGCCAAGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613200 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDS5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDS5A - PDS5 cohesin associated factor A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100399.1 | Intron | NP_001093869.1 | ||||
NM_001100400.1 | Intron | NP_001093870.1 | ||||
XM_011513672.2 | Intron | XP_011511974.1 | ||||
XM_017007928.1 | Intron | XP_016863417.1 |