Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAGTTCAGATGTGTTCCAGTCA[C/T]GTTCAGTGTTGTGTGACTGTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDS1 - CDP-diacylglycerol synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001263.3 | Intron | NP_001254.2 | ||||
XM_005262687.2 | Intron | XP_005262744.1 | ||||
XM_017007648.1 | Intron | XP_016863137.1 | ||||
XM_017007649.1 | Intron | XP_016863138.1 | ||||
XM_017007650.1 | Intron | XP_016863139.1 | ||||
XM_017007651.1 | Intron | XP_016863140.1 |