Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAATCCTCAAAACAAGGTCTAGC[C/T]AAGTTTATTGTTTGGTAATTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603262 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPSS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAPSS1 - 3'-phosphoadenosine 5'-phosphosulfate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005443.4 | 2408 | UTR 3 | NP_005434.4 | |||
XM_011532400.1 | 2408 | UTR 3 | XP_011530702.1 | |||
XM_011532401.1 | 2408 | UTR 3 | XP_011530703.1 |