Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTACATGCCTCTTAATTGGAT[A/G]CAATAGGTTCTGTATAGGAAATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM160A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM160A1 - family with sequence similarity 160 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109977.1 | Intron | NP_001103447.1 | ||||
XM_005263199.1 | Intron | XP_005263256.1 | ||||
XM_011532220.1 | Intron | XP_011530522.1 | ||||
XM_011532221.2 | Intron | XP_011530523.1 | ||||
XM_011532222.2 | Intron | XP_011530524.1 | ||||
XM_011532223.2 | Intron | XP_011530525.1 | ||||
XM_011532224.2 | Intron | XP_011530526.1 | ||||
XM_017008583.1 | Intron | XP_016864072.1 |