Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTATCCTTCAAACTAGCGGCTTG[A/G]TAAGAGTATCTCTTGCCCCCTTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985184 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107985184 - protein enabled homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017003082.1 | 25 | UTR 5 | XP_016858571.1 |
RAP1A - RAP1A, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010935.2 | 25 | Intron | NP_001010935.1 | |||
NM_001291896.1 | 25 | Intron | NP_001278825.1 | |||
NM_002884.3 | 25 | Intron | NP_002875.1 | |||
XM_017001960.1 | 25 | Intron | XP_016857449.1 | |||
XM_017001961.1 | 25 | Intron | XP_016857450.1 | |||
XM_017001962.1 | 25 | Intron | XP_016857451.1 | |||
XM_017001963.1 | 25 | Intron | XP_016857452.1 | |||
XM_017001964.1 | 25 | Intron | XP_016857453.1 |