Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTCTTTTACTGCAAAGACCAGC[A/G]CCGTGGGGGCCTGCGCTACATGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OBP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OBP2A - odorant binding protein 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293189.1 | Intron | NP_001280118.1 | ||||
NM_001293193.1 | Intron | NP_001280122.1 | ||||
NM_014582.2 | Intron | NP_055397.1 | ||||
XM_017014656.1 | Intron | XP_016870145.1 |