Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCCCGCCTTTTTATACTTTTGC[G/T]TGTGGAGGGCTTTTTGAACCGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPO - thymopoietin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032283.2 | Intron | NP_001027454.1 | ||||
NM_001032284.2 | Intron | NP_001027455.1 | ||||
NM_001307975.1 | Intron | NP_001294904.1 | ||||
NM_003276.2 | Intron | NP_003267.1 | ||||
XM_005269132.3 | Intron | XP_005269189.1 | ||||
XM_017019914.1 | Intron | XP_016875403.1 |
TMPO-AS1 - TMPO antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |