Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGACTTCCCTCAGACCCACCCC[C/T]ACAAGAGGCCATTCTGTGAATTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FICD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FICD - FIC domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SART3 - squamous cell carcinoma antigen recognized by T-cells 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014706.3 | 3117 | UTR 3 | NP_055521.1 | |||
XM_005269241.4 | 3117 | UTR 3 | XP_005269298.1 | |||
XM_011539026.2 | 3117 | UTR 3 | XP_011537328.1 |