Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGTGTTGCAAAAACTTAACTGC[C/T]CTCGGAACTTGCACGCGCCGTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALB - RALB Ras like proto-oncogene B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002881.2 | 54 | Intron | NP_002872.1 | |||
XM_005263724.1 | 54 | Intron | XP_005263781.1 | |||
XM_005263727.1 | 54 | Intron | XP_005263784.1 | |||
XM_005263728.1 | 54 | UTR 5 | XP_005263785.1 | |||
XM_005263729.2 | 54 | UTR 5 | XP_005263786.1 | |||
XM_011511573.1 | 54 | Intron | XP_011509875.1 | |||
XM_011511574.1 | 54 | Intron | XP_011509876.1 | |||
XM_017004621.1 | 54 | Intron | XP_016860110.1 | |||
XM_017004622.1 | 54 | Intron | XP_016860111.1 |