Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTCCTCCCTTCCTTGGCATCT[A/G]TGAGTGTGCAGCGTGTGGCTTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC5A7 - solute carrier family 5 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305005.2 | 22 | Intron | NP_001291934.1 | |||
NM_001305006.2 | 22 | Intron | NP_001291935.1 | |||
NM_001305007.2 | 22 | Intron | NP_001291936.1 | |||
NM_021815.4 | 22 | Intron | NP_068587.1 | |||
XM_011511580.2 | 22 | Intron | XP_011509882.1 | |||
XM_017004628.1 | 22 | Intron | XP_016860117.1 | |||
XM_017004629.1 | 22 | UTR 5 | XP_016860118.1 |