Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGAGGGCCGAGTCATTGAGTAA[C/T]GGGGCCTCTGCCCTCTCAGCCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601203 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL1RL1 - interleukin 1 receptor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282408.1 | Intron | NP_001269337.1 | ||||
NM_003856.3 | Intron | NP_003847.2 | ||||
NM_016232.4 | Intron | NP_057316.3 | ||||
XM_006712839.3 | Intron | XP_006712902.1 | ||||
XM_011512151.1 | Intron | XP_011510453.1 |