Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGTACTCCACTGGGAAGACCAC[C/T]TTCATCAGGTGAGCCGGCCCAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610229 MIM: 605891 | ||||||||||||||||||||
Literature Links: |
CAPN14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN14 - calpain 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145122.1 | 827 | Intron | NP_001138594.1 | |||
NM_001321270.1 | 827 | Intron | NP_001308199.1 | |||
XM_011532863.2 | 827 | Intron | XP_011531165.1 | |||
XM_011532864.2 | 827 | Intron | XP_011531166.1 | |||
XM_011532865.1 | 827 | Intron | XP_011531167.1 | |||
XM_011532866.2 | 827 | Intron | XP_011531168.1 |
EHD3 - EH domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014600.2 | 827 | Silent Mutation | ACC,ACT | T,T 73 | NP_055415.1 | |
XM_011532806.2 | 827 | Intron | XP_011531108.1 |