Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACACAATATGCCCCCAAACATG[A/G]ACCATTATTCAAGTAGACAAAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFPT1 - glutamine--fructose-6-phosphate transaminase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244710.1 | 5592 | UTR 3 | NP_001231639.1 | |||
NM_002056.3 | 5592 | UTR 3 | NP_002047.2 | |||
XM_017003801.1 | 5592 | UTR 3 | XP_016859290.1 | |||
XM_017003802.1 | 5592 | Intron | XP_016859291.1 |