Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAGGGAGAGGTCCAAAGAAGTT[A/G]AATGACTTCTCCAGAATCAAACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC142 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC142 - coiled-coil domain containing 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBX2 - ladybird homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC31 - tetratricopeptide repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022492.4 | Intron | NP_071937.4 | ||||
XM_005264492.1 | Intron | XP_005264549.1 | ||||
XM_011533035.1 | Intron | XP_011531337.1 | ||||
XM_011533036.1 | Intron | XP_011531338.1 | ||||
XM_011533037.1 | Intron | XP_011531339.1 | ||||
XM_011533040.1 | Intron | XP_011531342.1 | ||||
XM_017004706.1 | Intron | XP_016860195.1 | ||||
XM_017004707.1 | Intron | XP_016860196.1 | ||||
XM_017004708.1 | Intron | XP_016860197.1 | ||||
XM_017004709.1 | Intron | XP_016860198.1 |