Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTATCATTATCTCAGTTTTATA[C/G]ATGAGGAAATTAATGCACTGAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614719 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCMF1 - potassium channel modulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020122.4 | Intron | NP_064507.3 | ||||
XM_006712052.3 | Intron | XP_006712115.1 | ||||
XM_011532990.1 | Intron | XP_011531292.1 | ||||
XM_017004511.1 | Intron | XP_016860000.1 |