Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATGACCAAGAGTACCAAGTTTA[C/T]ATTAGTGTGTTTATCTACCATCAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608426 | |||||||||||||||||||||||
Literature Links: |
MKRN2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MKRN2 - makorin ring finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271707.1 | Intron | NP_001258636.1 | ||||
NM_014160.4 | Intron | NP_054879.3 |
MKRN2OS - MKRN2 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195279.1 | Intron | NP_001182208.1 | ||||
XM_006712909.3 | Intron | XP_006712972.1 | ||||
XM_011533241.2 | Intron | XP_011531543.1 | ||||
XM_011533242.2 | Intron | XP_011531544.1 | ||||
XM_011533243.2 | Intron | XP_011531545.1 |