Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGTTCCTGGAAGGAGGTGGGA[A/G]CAGTAGCTCTGGAAGAGGGCTGTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607416 | |||||||||||||||||||||||
Literature Links: |
CHL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CHL1 - cell adhesion molecule L1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253387.1 | Intron | NP_001240316.1 | ||||
NM_001253388.1 | Intron | NP_001240317.1 | ||||
NM_006614.3 | Intron | NP_006605.2 | ||||
XM_006712938.1 | Intron | XP_006713001.1 | ||||
XM_006712939.3 | Intron | XP_006713002.1 | ||||
XM_006712940.3 | Intron | XP_006713003.1 | ||||
XM_011533292.1 | Intron | XP_011531594.1 | ||||
XM_011533294.1 | Intron | XP_011531596.1 | ||||
XM_011533295.1 | Intron | XP_011531597.1 | ||||
XM_011533296.1 | Intron | XP_011531598.1 | ||||
XM_017005566.1 | Intron | XP_016861055.1 | ||||
XM_017005567.1 | Intron | XP_016861056.1 | ||||
XM_017005568.1 | Intron | XP_016861057.1 | ||||
XM_017005569.1 | Intron | XP_016861058.1 | ||||
XM_017005570.1 | Intron | XP_016861059.1 | ||||
XM_017005571.1 | Intron | XP_016861060.1 | ||||
XM_017005572.1 | Intron | XP_016861061.1 | ||||
XM_017005573.1 | Intron | XP_016861062.1 |
LOC101927174 - uncharacterized LOC101927174 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |