Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCATCTCTATGGCTAAGAAAA[C/T]GCCAGTTAGTGAAACACGATGGACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603353 | |||||||||||||||||||||||
Literature Links: |
NEK10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NEK10 - NIMA related kinase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC4A7 - solute carrier family 4 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258379.1 | 3868 | UTR 3 | NP_001245308.1 | |||
NM_001258380.1 | 3868 | UTR 3 | NP_001245309.1 | |||
NM_001321103.1 | 3868 | UTR 3 | NP_001308032.1 | |||
NM_001321104.1 | 3868 | UTR 3 | NP_001308033.1 | |||
NM_001321105.1 | 3868 | UTR 3 | NP_001308034.1 | |||
NM_001321106.1 | 3868 | UTR 3 | NP_001308035.1 | |||
NM_001321107.1 | 3868 | UTR 3 | NP_001308036.1 | |||
NM_001321108.1 | 3868 | UTR 3 | NP_001308037.1 | |||
NM_003615.4 | 3868 | UTR 3 | NP_003606.3 | |||
XM_005265598.4 | 3868 | UTR 3 | XP_005265655.1 | |||
XM_005265600.4 | 3868 | UTR 3 | XP_005265657.1 | |||
XM_006713421.2 | 3868 | UTR 3 | XP_006713484.1 | |||
XM_011534256.2 | 3868 | UTR 3 | XP_011532558.1 | |||
XM_011534258.1 | 3868 | UTR 3 | XP_011532560.1 | |||
XM_011534261.2 | 3868 | UTR 3 | XP_011532563.1 | |||
XM_011534263.2 | 3868 | UTR 3 | XP_011532565.1 | |||
XM_011534265.1 | 3868 | UTR 3 | XP_011532567.1 | |||
XM_017007526.1 | 3868 | UTR 3 | XP_016863015.1 | |||
XM_017007527.1 | 3868 | UTR 3 | XP_016863016.1 | |||
XM_017007528.1 | 3868 | UTR 3 | XP_016863017.1 | |||
XM_017007529.1 | 3868 | UTR 3 | XP_016863018.1 |