Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTGGTGTTCATCTTTGGTTTT[A/G]TGGGCAACATGCTGGTCATCCTCAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601373 | |||||||||||||||||||||||
Literature Links: |
CCR5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCR5 - C-C motif chemokine receptor 5 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000579.3 | 493 | Missense Mutation | ATG,GTG | M,V 46 | NP_000570.1 | |
NM_001100168.1 | 493 | Missense Mutation | ATG,GTG | M,V 46 | NP_001093638.1 |
LOC102724297 - uncharacterized LOC102724297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |