Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTACCCTCCTAACTTATCTA[C/G]GTGCCAGTGACTTAACTAGGTCCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606525 | |||||||||||||||||||||||
Literature Links: |
SRGAP3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SRGAP3 - SLIT-ROBO Rho GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033117.2 | 6097 | UTR 3 | NP_001028289.1 | |||
NM_014850.3 | 6097 | UTR 3 | NP_055665.1 | |||
XM_011534295.1 | 6097 | UTR 3 | XP_011532597.1 | |||
XM_011534296.1 | 6097 | UTR 3 | XP_011532598.1 | |||
XM_011534301.2 | 6097 | UTR 3 | XP_011532603.1 | |||
XM_011534303.1 | 6097 | UTR 3 | XP_011532605.1 | |||
XM_017007574.1 | 6097 | UTR 3 | XP_016863063.1 | |||
XM_017007575.1 | 6097 | UTR 3 | XP_016863064.1 | |||
XM_017007576.1 | 6097 | UTR 3 | XP_016863065.1 | |||
XM_017007577.1 | 6097 | UTR 3 | XP_016863066.1 | |||
XM_017007578.1 | 6097 | UTR 3 | XP_016863067.1 | |||
XM_017007579.1 | 6097 | UTR 3 | XP_016863068.1 | |||
XM_017007580.1 | 6097 | Intron | XP_016863069.1 |