Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGATCCCCATTTTTTAGAACA[A/T]TGAGTGGGCACACGATGGCCCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614572 | ||||||||||||||||||||
Literature Links: |
ZFP42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP42 - ZFP42 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304358.1 | Intron | NP_001291287.1 | ||||
NM_174900.4 | Intron | NP_777560.2 | ||||
XM_011531604.2 | Intron | XP_011529906.1 | ||||
XM_011531605.1 | Intron | XP_011529907.1 | ||||
XM_011531606.1 | Intron | XP_011529908.1 | ||||
XM_011531607.2 | Intron | XP_011529909.1 |