Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCGCGCGGCTTGGGCGGACGT[A/C]TCGTGAGACGTGGGACTTCTCGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPDL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPDL1 - spindle apparatus coiled-coil protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017785.4 | 32 | UTR 5 | NP_060255.3 | |||
XM_011534585.1 | 32 | UTR 5 | XP_011532887.1 | |||
XM_011534586.2 | 32 | Intron | XP_011532888.1 | |||
XM_011534587.2 | 32 | UTR 5 | XP_011532889.1 | |||
XM_011534588.2 | 32 | UTR 5 | XP_011532890.1 | |||
XM_011534589.2 | 32 | Intron | XP_011532891.1 | |||
XM_011534590.2 | 32 | Intron | XP_011532892.1 | |||
XM_017009601.1 | 32 | Intron | XP_016865090.1 | |||
XM_017009602.1 | 32 | Intron | XP_016865091.1 |