Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCAGCGGGTCTGGCCCAGTGTA[C/T]GTTGTGTGGCATGTGTGGGTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNIP1 - potassium voltage-gated channel interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034837.2 | Intron | NP_001030009.1 | ||||
NM_001034838.2 | Intron | NP_001030010.1 | ||||
NM_001278339.1 | Intron | NP_001265268.1 | ||||
NM_001278340.1 | Intron | NP_001265269.1 | ||||
NM_014592.3 | Intron | NP_055407.1 | ||||
XM_017009407.1 | Intron | XP_016864896.1 | ||||
XM_017009408.1 | Intron | XP_016864897.1 |